Would you like to log in again?
Bottle top vacuum filtration systems, High Performance
Supplier: VWR Collection
Designed for the preparation of buffers, tissue culture media, microbiological media and other biological fluids. These vacuum filtration systems feature large diameter Pall High Performance PES membranes for guaranteed performance and maximum throughput. The full assembly includes a filtration unit and a receiver bottle with an ergonomic shape, moulded finger grips and a patented bottle cap. The centrifuge tube and funnel system allows the user to filter fluids directly into a 15 or 50 ml tube.
  • Quick-Connect™ pedestal base or cradle ring allows for hands-free filtration. Weighted for extra stability
  • Quick-Connect™ pedestal base ensures that the unit is always correctly placed
  • Forward-facing, large, raised graduation marks allow for easy visualisation of volumes
  • Vacuum on/off switch located at the front
  • Vacuum hose adapter to use with conventional filtration set-ups
Each separate piece is compatible with the Quick-Connect™ pedestal base providing a convenient on/off vacuum flow control, which allows for a convenient one-time vacuum hose connection and one handed operation. A cradle ring is also available for use with existing laboratory stands. Systems are available with 0,1 µm (mycoplasma filtering grade), 0,2 µm (sterilising grade) or 0,45 µm (clarification grade) asymmetric PES membranes. All items are made from FDA grade, Class VI materials and are gamma sterilised VD Max 10-6 in a 100,000 cleanroom in ISO 13485 standard facility.
You can choose between complete systems and filtration funnels only. For the complete systems there is a choice of receivers: 15 or 50 ml centrifuge tubes; 250, 500 or 1000 ml media bottles.
Delivery information: Complete system includes filtration funnel (PS), media bottle (PS) or centrifuge tube (PP) and hose adapter.
Log in to see your contract pricing and availability.
  Remember me on this device.
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Complete filtration units
Description Membrane Capacity Pore size Membrane Ø Pk VWR Catalog Number Quantity
Centrifuge tube filtration unit PES 15 ml 0,1 µm 40 mm 24 514-0367
Centrifuge tube filtration unit PES 15 ml 0,2 µm 40 mm 24 514-0317
Centrifuge tube filtration unit PES 15 ml 0,45 µm 40 mm 24 514-0318
Centrifuge tube filtration unit PES 50 ml 0,1 µm 40 mm 24 514-0366
Centrifuge tube filtration unit PES 50 ml 0,2 µm 40 mm 24 514-0308
Centrifuge tube filtration unit PES 50 ml 0,45 µm 40 mm 24 514-0309
Media bottle filtration unit PES 250 ml 0,1 µm 66 mm 12 514-0348
Media bottle filtration unit PES 250 ml 0,2 µm 66 mm 12 514-0296
Media bottle filtration unit PES 250 ml 0,45 µm 66 mm 12 514-0299
Media bottle filtration unit PES 500 ml 0,1 µm 90 mm 12 514-0349
Media bottle filtration unit PES 500 ml 0,2 µm 90 mm 12 514-0297
Media bottle filtration unit PES 500 ml 0,45 µm 90 mm 12 514-0300
Media bottle filtration unit PES 1000 ml 0,1 µm 90 mm 12 514-0350
Media bottle filtration unit PES 1000 ml 0,2 µm 90 mm 12 514-0298
Media bottle filtration unit PES 1000 ml 0,45 µm 90 mm 12 514-0301
Filtration funnels only
Description Membrane Capacity Pore size Membrane Ø Pk VWR Catalog Number Quantity
Filtration funnel for centrifuge tube PES 15 ml 0,1 µm 40 mm 48 514-0363
Filtration funnel for centrifuge tube PES 15 ml 0,2 µm 40 mm 48 514-0364
Filtration funnel for centrifuge tube PES 15 ml 0,45 µm 40 mm 48 514-0365
Filtration funnel for centrifuge tube PES 50 ml 0,1 µm 40 mm 48 514-0359
Filtration funnel for centrifuge tube PES 50 ml 0,2 µm 40 mm 48 514-0360
Filtration funnel for centrifuge tube PES 50 ml 0,45 µm 40 mm 48 514-0362
Filtration funnel for media bottle PES 250 ml 0,1 µm 66 mm 24 514-0354
Filtration funnel for media bottle PES 250 ml 0,2 µm 66 mm 24 514-0302
Filtration funnel for media bottle PES 250 ml 0,45 µm 66 mm 24 514-0305
Filtration funnel for media bottle PES 500 ml 0,1 µm 90 mm 24 514-0355
Filtration funnel for media bottle PES 500 ml 0,2 µm 90 mm 24 514-0303
Filtration funnel for media bottle PES 500 ml 0,45 µm 90 mm 24 514-0306
Filtration funnel for media bottle PES 1000 ml 0,1 µm 90 mm 24 514-0358
Filtration funnel for media bottle PES 1000 ml 0,2 µm 90 mm 24 514-0304
Filtration funnel for media bottle PES 1000 ml 0,45 µm 90 mm 24 514-0307
PS bottles with PP cap, sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0286
250 ml 24 391-0283
500 ml 24 391-0284
PC bottles with PP cap, sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0299
250 ml 24 391-0297
500 ml 24 391-0298
PETG bottles with PP cap, sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0296
250 ml 24 391-0294
500 ml 24 391-0295
PS bottles with PP cap, non sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0319
250 ml 24 391-0317
PC bottles with PP cap, non sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0323
250 ml 24 391-0321
500 ml 24 391-0322
PETG bottles with PP cap, non sterile
Capacity Pk VWR Catalog Number Quantity
1000 ml 24 391-0316
250 ml 24 391-0314
500 ml 24 391-0315
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Cradle ring 1 514-0324
Quick-Connect™ pedestal base 1 514-0319
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
???store.checkout.costsavings.reviewAndCompare???
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty