Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "shirts"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/97/90/18989790.jpg
SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Catalog Number: (KWIN100222530XS)
Supplier:  FRISTADS KANSAS GROUP
Description:   Heavy men's piqué poloshirt made of 100% pre-shrunk, ring-spun and combed cotton. The model in grey melange is made of 90% cotton and 10% viscose.
UOM:  1 * 1 items
1 - 1  of 1