  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Tubing for Circulators"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/99/48/18989948.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"15"
Sort by:
Supplier:  VWR Collection
Description:   Tubing for circulator, VWR®, Length: 1000 mm, Ø int.: 6 mm, Buran N (synthetic rubber) tubing
Catalog Number: (462-0300)
Supplier:  VWR Collection
Description:   Fluoroelastomer, black.
UOM:  1 * 1 items
Supplier:  JULABO GMBH
Description:   Tubing for circulator, Viton® tubing, working temperature -35 to +200 °C, Ø int.: 12 mm
Supplier:  JULABO GMBH
Description:   Metal tubing is integrated insulated and flexible with connectors and fittings. This flexible metal tubing can be bolted together with the circulator ...
Supplier:  JULABO GMBH
Description:   Working temperature -40 to +120 °C.
Supplier:  GRANT INSTRUMENTS LTD.
Description:   Tubing kit 2 x 2m, assembled with OptimaTM pump outlet plate and simple hose clips, no tools required (can be cut to length as required)
Supplier:  LAUDA
Description:   This insulated tubing is used with thermostats.
Supplier:  LAUDA
Description:   These metal hoses with thermal insulation consist of flexible corrugated stainless steel tubing and are unaffected by the usual bath liquids. The hose...
1 - 8  of 15
  1  2  Next