  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Cryogenic Envelopes"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  TENAK A/S
Description:   Bloodbag envelope, Cardboard, White, Volume: 500 ml, W×D×H: 235×135×17 mm
1 - 1  of 1