  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "GC Sorbents"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Catalog Number: (HYPETS-49350)
Supplier:  THERMOQUEST HYPERSIL
Description:   Alkylation reagents
UOM:  1 * 25 ml
1 - 1  of 1