  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Water Bath Protection Reagents"
https://tr.vwr.com/stibo/smallweb/std.lang.all/00/12/18990012.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  VWR Chemicals
Description:   Water bath protective agent, Aquaresist
UOM:  1 * 100 ml
SDS
Supplier:  PanReac AppliChem
Description:   Water bath disinfection reagent, Aquabator-Clean™
UOM:  1 * 250 ml
Supplier:  JULABO GMBH
Description:   Water bath protective media, Aqua-Stabil
Supplier:  SPECTRUM LABORATORIES
Description:   Water bath treatment, Clear Bath®
Supplier:  GFL (GESELLSCH. FÜR LABORT.)
Description:   Water Bath Protection Agent, ProAquaTop
1 - 5  of 5