  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Redox/ORP Standards"
https://tr.vwr.com/stibo/smallweb/std.lang.all/97/21/18989721.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Supplier:  EHRENSTORFER GMBH
Description:   Organic reference standard, o-Dianisidine 100 µg/ml in acetonitrile
UOM:  1 * 1 ml
Supplier:  HANNA
Description:   Redox solutions, Hanna
Supplier:  THERMO ORION
Description:   ORP standards for Redox/ORP electrodes, Orion™
1 - 3  of 3