  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Filter Tiles"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/68/99/20236899.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  BOHLENDER
Description:   Filter tiles, PTFE, Dimensions: 320×320 mm, Pore size: 10 μm, Thickness: 2 mm
1 - 1  of 1