Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Electrolytes"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/51/79/18945179.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"35"
Sort by:
Supplier:  VWR Chemicals
Description:   Potassium chloride 3M solution may also be used as an electrode storage solution.
SDS
Supplier:  VWR Collection
Description:   Refill solution 3,5 mol/l KCl, VWR®
UOM:  1 * 100 ml
SDS
Catalog Number: (662-1116)
Supplier:  VWR Collection
Description:   Calomel-free pH electrode refill solution containing potassium nitrate
UOM:  1 * 114 ml
Catalog Number: (705-1030)
Supplier:  Hach
Description:   Potassium chloride (KCl) 3 M
UOM:  1 * 500 ml
Supplier:  THERMO ORION
Description:   Electrode storage solution for ammonia ion selective electrode, 475 ml bottle, Orion
UOM:  1 * 1 items
Supplier:  Hach
Description:   Chloride standard solution, 12,500 mg/l as Cl- (NIST), 10 ml Voluette® ampules
Supplier:  Hach
Description:   Electrolyte filling solution (50 ml) for sensION and sensION+ 5130 dissolved oxygen probes
Supplier:  Hach
Description:   Internal filling solution for use with PHC281 IntelliCAL™ Ultra pH probes, 2,44M KCl, 59 ml bottle with dispenser cap, IntelliCAL™
UOM:  1 * 59 ml
1 - 8  of 35
  1  2  3  4  5  Next