Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Protein Reagents"
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"37"
Sort by:
Supplier:  VWR Chemicals
Description:   Cytoplasmic/Nuclear Protein Enrichment Kit
UOM:  1 * 1 KIT
Catalog Number: (M329-10ML)
Supplier:  VWR Chemicals
Description:   Total Protein Cell Lysis Buffer
UOM:  1 * 10 ml
Catalog Number: (N990-1L)
Supplier:  VWR Chemicals
Description:   Autoinduction media, ZYP-5052
UOM:  1 * 1 l
Catalog Number: (1B1622-KIT)
Supplier:  VWR Chemicals
Description:   Fluorescent Protein Quantitation Kit
UOM:  1 * 1 KIT
Supplier:  GE Healthcare - Life Sciences
Description:   Gel filtration media, Sephadex™ G-50
Supplier:  Protea Biosciences
Description:   MALDI matrix, ultrapure sinapic acid
Supplier:  Protea Biosciences
Description:   MALDI matrix, ultrapure 3-HPA
Catalog Number: (PTEASP-817-10KIT)
Supplier:  Protea Biosciences
Description:   ProteaPrep™ Non-ionic Cell Lysis Kit
UOM:  1 * 1 items
1 - 8  of 37
  1  2  3  4  5  Next