  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Trypsinizing Flasks"
https://tr.vwr.com/stibo/smallweb/std.lang.all/53/56/18945356.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Supplier:  WHEATON INDUSTRIES INC
Description:   Trypsinizing flasks, non graduated, with pourout and screw caps, Capacity: 75 ml, Top screw cap 33-430, Neck: Screw cap 20-415
Supplier:  WHEATON INDUSTRIES INC
Description:   These trypsinizing flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improv...
Supplier:  WITEG LABORTECHNIK GMBH
Description:   Trypsinizing flasks, graduated, designed for converting tissue fragments and homogenates into
cell suspensions by trypsin digestion of connective...
1 - 3  of 3