  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Microplates with Glass Inserts"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/46/18989546.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
Sort by:
Supplier:  VWR Collection
Description:   96-Position systems (Ritter) with glass inserts are used in HTS (high throughput screening) chemistry when the pure PP block is not inert enough. The ...
Supplier:  VWR Collection
Description:   High quality polypropylene microplates coated with 200 nm thick layer of silicone dioxide.
Supplier:  VWR Collection
Description:   High quality polypropylene microplates coated with 200 nm thick layer of silicone dioxide.
Supplier:  CHROMACOL
Description:   Convenient 96-well plate kits with pre-inserted glass or PTFE vials. Compatible with autosamplers that have 96-well plate configuration.
Supplier:  CHROMACOL
Description:   High quality polypropylene microplates coated with 200 nm thick layer of silicone dioxide.
Catalog Number: (391-1972)
Supplier:  HIRSCHMANN
Description:   Microplate, untreated, with fixed glass inserts, ~50 µl, No. of wells: 1536, PP, Microplate, PP, 1536-well, flat bottom with fixed glass inserts, chemically inert
UOM:  1 * 1 items
Supplier:  HIRSCHMANN
Description:   Highly pure, chemically inert aluminium plate for individual glass inserts. Translucent plastic cover included.
Supplier:  HIRSCHMANN
Description:   Microplate, untreated, with fixed glass inserts, ~1200 µl, No. of wells: 96, PP, Microplate, PP, 96-well with fixed glass inserts, chemically inert
1 - 8  of 8