  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Microplate Heat Sealers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/42/18989542.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Catalog Number: (732-0996)
Supplier:  MATRIX TECHNOLOGIES CORP. LTD
Description:   Microplate heat sealer, ALPS™ 50V
UOM:  1 * 1 items
Supplier:  AXYGEN
Description:   Semi automated microplate heat sealer, Axygen® PlateMax®
UOM:  1 * 1 items
Supplier:  EPPENDORF
Description:   Heat sealers, Eppendorf HeatSealer S100/S200
1 - 3  of 3