  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Halogen Lamps"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/81/18989381.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Supplier:  VWR Collection
Description:   Halogen bulb (6 V/30 W), 6 V/30 W
Catalog Number: (631-0518)
Supplier:  SCHOTT AG (LIGHTING AND IMAGIN
Description:   Halogen bulb, 15 V / 150 W, 15V/150W
UOM:  1 * 1 items
Supplier:  MOTIC
Description:   Quartz halogen lamp, 6 V/30 W, 6V/30W
1 - 3  of 3