  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Weather Stations"
https://tr.vwr.com/stibo/smallweb/std.lang.all/00/21/18990021.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"9"
Sort by:
Catalog Number: (620-0906)
Supplier:  VWR Collection
Description:   This weather station is designed for use in agriculture, offices and universities. It includes an internal temperature sensor.
UOM:  1 * 1 items
Catalog Number: (620-2120)
Supplier:  VWR Collection
Description:   The Cumulus weather station is designed for use in agriculture, offices and universities. Snooze alarm, moon phase display, dual scale thermometer (°C...
UOM:  1 * 1 items
Catalog Number: (620-2127)
Supplier:  VWR Collection
Description:   Calendar up to the year 2050 with date and day of week in seven languages (English, German, French, Italian, Spanish, Dutch and Danish), a 12/24-hour ...
UOM:  1 * 1 items
Catalog Number: (620-2128)
Supplier:  VWR Collection
Description:   This weather station features a perpetual calendar up to year 2099 with date, 12/24 hour selectable time with snooze alarm, moon phase, tide indicator...
UOM:  1 * 1 items
Supplier:  TESTO
Description:   The testo 435 measuring instruments are designed to analyse the ventilation and indoor air quality of the working environment, serving as an indicator...
Catalog Number: (620-1105)
Supplier:  AMARELL, ARNO
Description:   Rain gauge acc. to Prof. Hellmann, 0 - 10000 mm²
UOM:  1 * 1 items
Supplier:  TESTO
Description:   This measuring instrument is designed to analyse the ventilation and indoor air quality of the working environment, serving as an indicator for the we...
UOM:  1 * 1 items
Supplier:  TESTO
Description:   This portable instrument measures temperature, relative humidity, dew point, absolute humidity, degree of humidity, enthalpy, all types of air velocit...
UOM:  1 * 1 items
1 - 8  of 9
  1  2  Next