Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Spectrophotometers and Spectrometers"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/98/24/18989824.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"32"
Sort by:
Supplier:  VWR Collection
Description:   mySPEC allows to perform nucleic acids and proteins microvolume concentration and purity measurements.
Catalog Number: (634-6040)
Supplier:  VWR Collection
Description:   Spectrophotometer for advanced routine measurements and education applications.
UOM:  1 * 1 items
Catalog Number: (634-6042)
Supplier:  VWR Collection
Description:   Spectrophotometer for advanced measurements in quality control, biochemical and pharmaceutical applcaitions.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   The V-1200 is designed for routine daily quality control requirements, in-process control and teaching laboratories.
Catalog Number: (634-6041)
Supplier:  VWR Collection
Description:   A double beam spectrophotometer suitable for a wide range of applications including general research, pharmaceutical, biochemical and clinical laborat...
UOM:  1 * 1 items
Supplier:  WTW
Description:   The pHotoLab® 7100 Vis and 7600 UV-Vis combine routine analysis with spectral analysis and pioneering procedure OptRF.
Supplier:  WTW
Description:   Filter photometer with six wavelengths for common routine testing of wastewater and drinking water using cell tests (Ø 16 mm round cells).
Catalog Number: (634-0757)
Supplier:  VWR Collection
Description:   The U-5100 can be used in a wide range of quantitative analytical applications that includes biotechnology or quality control laboratories.
UOM:  1 * 1 items
1 - 8  of 32
  1  2  3  4  Next