  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Mills"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/51/18989551.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"37"
Sort by:
Catalog Number: (412-0167)
Supplier:  VWR Collection
Description:   A ball mill for disintregration of small sample volumes that can also be used to shake/mix microtubes and microplates. Wide range of applications in d...
UOM:  1 * 1 items
Catalog Number: (42627.KT)
Supplier:  ALFA (AESAR)
Description:   These gringing mills are ideal for mixing and grinding tasks that require constant or long periods of operation to mix the sample for IR analysis.
UOM:  1 * 1 items
Catalog Number: (412-1111)
Supplier:  IKA (JANKE & KUNKEL )
Description:   This batch mill performs two different types of grinding; impact grinding of hard, brittle or non-elastic materials by means of the high-grade stainle...
UOM:  1 * 1 items
Catalog Number: (412-0203)
Supplier:  IKA (JANKE & KUNKEL )
Description:   The Tube Mill control is a highly versatile small batch mill with disposable grinding chambers thereby eliminating the possibility of cross-contaminat...
UOM:  1 * 1 items
Supplier:  MORGAN ADVANCED MATERIALS HALD
Description:   Mill, Ball mill, GSK, 1700 ml, base Ø 150 mm, height 222 mm
Catalog Number: (412-1130)
Supplier:  IKA (JANKE & KUNKEL )
Description:   Continuously operating universal grinder with powerful drive. When using the grinding heads, the processed material passes through an interchangeable ...
UOM:  1 * 1 items
Catalog Number: (412-1120)
Supplier:  IKA (JANKE & KUNKEL )
Description:   This batch mill with stainless steel chamber is suitable for dry grinding of hard and brittle substances.
UOM:  1 * 1 items
Catalog Number: (RETS20.510.0001)
Supplier:  RETSCH GMBH
Description:   This high energy ball mill is ideal for nano grinding, size reduction, homogenising, mechanical alloying, colloidal milling and high energy comminutio...
UOM:  1 * 1 items
1 - 8  of 37
  1  2  3  4  5  Next