  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Microplate Dispensers"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/95/39/18989539.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Catalog Number: (735-0441)
Supplier:  LABSYSTEMS LIFE SCIENCE
Description:   The Multidrop® DW is a high-speed, automated dispenser designed for Deep Well plates, 1,1ml test tubes and 96-well plates. The instrument dispenses vo...
UOM:  1 * 1 items
Supplier:  LABSYSTEMS LIFE SCIENCE
Description:   Based on the proven and reliable Multidrop® range, the Multidrop® Combi provides high speed bulk reagent dispensing for biotech and pharmaceutical lab...
Catalog Number: (735-0440)
Supplier:  LABSYSTEMS LIFE SCIENCE
Description:   For today's pharmaceutical and research laboratories, 384-well plates offer a number of important benefits, including lower consumption of reagents an...
UOM:  1 * 1 items
1 - 3  of 3