Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Hygrometers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/94/13/18989413.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"48"
Sort by:
Supplier:  VWR Collection
Description:   This digital thermo-hygrometer continuously displays both relative humidity and temperature.
UOM:  1 * 1 items
Catalog Number: (620-1952)
Supplier:  VWR Collection
Description:   This accurate thermo-hygrometer with barometer features additional derived units: Dew point temperature, wet bulb temperature, moisture content and ab...
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Digital thermo-hygrometer with probe, Traceable®, −40…+104 °C; 10 - 95 % RH , 89×140×32 mm
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   These thermo-hygrometers are ideal for routine measurements, monitoring round-the-clock, all quality-control needs, and critical experimental requirem...
Supplier:  VWR Collection
Description:   Convenient triple reading meter with 'always on' feature for continuous monitoring conditions. Audible and LED alarm may be set to trigger at three di...
UOM:  1 * 1 items
Catalog Number: (628-0021)
Supplier:  VWR Collection
Description:   This thermo-hygrometer provides instant readings for constant monitoring labs, fume hoods, walk-ins, plant areas, and storage facilities.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Digital thermo-hygrometer, Traceable®, 0…+50 °C (in); –50…+70 °C (ex); 25 - 95% rF, 108×19×107 mm
UOM:  1 * 1 items
Catalog Number: (620-0923)
Supplier:  VWR Collection
Description:   For all environments, this analogue thermo-hygrometer can withstand use inside incubators, refrigerators and freezers. It features contrasting black t...
UOM:  1 * 1 items
1 - 8  of 48
  1  2  3  4  5  6  Next