  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Weighing Tables"
https://tr.vwr.com/stibo/smallweb/std.lang.all/00/24/18990024.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Catalog Number: (611-2536)
Supplier:  VWR Collection
Description:   Weighing table, Anti-vibration weighing surface, granite, W×D×H: 560×450×65 mm
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   High performance microbalances and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast st...
UOM:  1 * 1 items
1 - 2  of 2