  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Storage Cabinets"
https://tr.vwr.com/stibo/smallweb/std.lang.all/98/63/18989863.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"95"
Sort by:
Supplier:  DÜPERTHAL GMBH & CO
Description:   Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Ventilated Cabinet with filtration, Schoolcap AVPS 802
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Filtration system for safety cabinets, ChemTrap™
Catalog Number: (135-1451)
Supplier:  DÜPERTHAL GMBH & CO
Description:   Hazardous substances centre
UOM:  1 * 1 items
Supplier:  DÜPERTHAL GMBH & CO
Description:   Acids and lye materials storage cabinet, ACID Line
Supplier:  DÜPERTHAL GMBH & CO
Description:   Safety underbench cabinets, type 90, UTS ergo line S, 600 mm depth
Supplier:  DÜPERTHAL GMBH & CO
Description:   Safety underbench cabinets, type 90, UTS ergo line ST, 600 mm depth
Supplier:  DÜPERTHAL GMBH & CO
Description:   Safety underbench cabinets, type 90, UTS ergo line LD, 600 mm depth
1 - 8  of 95
  1  2  3  4  5  6  7  8  9  10  11  12  Next