  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Dry Ice Makers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/51/58/18945158.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  BÜRKLE
Description:   SnowPack enables the user to obtain a -79° C dry ice tablet within minutes, without complicated technology and without electricity. Can be installed o...
Supplier:  Bel-Art
Description:   Frigimat® dry ice makers produces ice flakes or snow form, which is used to pack irregular shaped items. A brass nozzle screws on the LCO₂ cylinder is...
Catalog Number: (VWRMNP560)
Supplier:  VWR BRAND (VWR-PRODUCTS USA)
Description:   These dry ice machines are ideal for tissue preservation, specimen shipping, biologicals manufacturing, industrial processes, ice cream, food samples,...
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Dry ice maker made of epoxy-coated steel with an aluminum ice chamber and brass valves with four non-skid rubber feet. Includes a 1,8 m nickel plated ...
Supplier:  Bel-Art
Description:   In these dry ice makers, the epoxy-coated steel frame has four non-skid rubber feet to prevent sliding.
1 - 5  of 5