  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Decappers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/40/40/18374040.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
Sort by:
Supplier:  VWR Collection
Description:   VWR®, Decapper for 8 mm aluminium caps
Supplier:  VWR Collection
Description:   VWR®, Decapper for 20 mm crimp caps
Supplier:  CHROMACOL
Description:   Chromacol™, Pliers type decapper for 13 mm crimp caps
Supplier:  CHROMACOL
Description:   Electronic decapper for 11 mm crimp caps, Pressure adjustable
Supplier:  BOCHEM
Description:   Decapper for 13 mm crimp caps
Supplier:  DWK Life Sciences (WHEATON®)
Description:   E-Z Decappers™, Decapper for 20 mm crimp caps
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Pliers type decapper for 13 mm seals
Catalog Number: (WHEA224357)
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Decapper for 30 mm aluminium caps
UOM:  1 * 1 items
1 - 8  of 8