  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Test/Sample Tubes"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/99/09/18989909.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"95"
Sort by:
Supplier:  VWR Collection
Description:   FIOLAX®, neutral glass.
Supplier:  VWR Collection
Description:   Soda-lime glass.
Supplier:  VWR Collection
Description:   Culture and test tube, VWR®, with stoppers, Conical bottom, PP, Capacity: 4 ml, Length: 75 mm, Ø ext.: 12 mm, Colour: Natural, PP, Packed: 25/Bag
Supplier:  VWR Collection
Description:   Culture, sample and test tubes, non-sterile, VWR®, without stoppers, Round bottom, PS, Capacity: 8 ml, Length: 100 mm, Ø ext.: 13 mm, PS, Packed: 1000/Bag
Supplier:  VWR Collection
Description:   Borosilicate glass, grade 3.3.
Supplier:  VWR Collection
Description:   AR-Glas®, soda lime glass.
Supplier:  VWR Collection
Description:   Polystyrene (PS), transparent or polypropylene (PP), translucent.
Supplier:  VWR Collection
Description:   Tubes, non sterile, VWR®, without rim and stopper, Round bottom, PS, Capacity: 5 ml, Length: 74,5 mm, Ø ext.: 13,0 mm, PS, Ø int.: 11,0 mm
1 - 8  of 95
  1  2  3  4  5  6  7  8  9  10  11  12  Next