  • Criteria
    • In Stock (5)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Microcentrifuge Tubes"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/31/18989531.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"76"
Sort by:
Supplier:  VWR Chemicals
Description:   Microcentrifuge tubes, DNase-/RNase-free
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 0,65 ml, SuperClear™ boil-proof
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 1,5 ml, SuperSpin™
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 1,7 ml, SuperClear™ boil-proof
UOM:  1 * 500 items
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 2,0 ml, SuperClear™
Supplier:  VWR Collection
Description:   Microcentrifuge tubes and screw caps
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 0,5/1,5/2,0 ml, SuperClear™
Supplier:  VWR Collection
Description:   Microcentrifuge tubes, 1,7 ml, SuperClear™
1 - 8  of 76
  1  2  3  4  5  6  7  8  9  10  Next