  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "immunology-tubes"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/94/27/18989427.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Supplier:  NUNC (INC)
Description:   Immunology tube, Nunc™, Length: 70 mm, Ø (mm): 11, Capacity: 5 ml, MiniSorp™, Ø ext.×H: 11×70 mm
Supplier:  NUNC (INC)
Description:   Immunology tube, Nunc™, Length: 75 mm, Ø (mm): 12, Capacity: 5,0 ml, Nunc StarTubes™, MaxiSorp™, Ø ext.×H: 12×75 mm
1 - 2  of 2