  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Flask Supports"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/68/18709568.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"11"
Sort by:
Supplier:  VWR Collection
Description:   Made of natural cork
Supplier:  VWR Collection
Description:   The ideal rest for round bottom flasks and other round bottom glass vessels. Compressed cork, natural.
Supplier:  VERNERET (INCL. NORTON) (PART
Description:   Can be used to provide a stable support for round bottom flasks and other containers with curved bases. Reversible. Non-slip, soft, stackable and easy...
Catalog Number: (IKAA4448400)
Supplier:  IKA (JANKE & KUNKEL )
Description:   This flask carrier with handle is constructed from anodised aluminium.
UOM:  1 * 1 items
Catalog Number: (214-0195)
Supplier:  USBECK LABORGERÄTE KG
Description:   Flask holder, Cork, For: 500 - 1000 ml, Ø ext.: 140 mm, I-Ø bottom: 90 mm, Height: 30 mm
UOM:  1 * 1 items
Catalog Number: (214-1920)
Supplier:  KARTELL
Description:   Provide secure, non-slip support for round bottom flasks and any other flasks with convex bases of any height. The concentric inner rings can hold fla...
UOM:  1 * 1 items
Catalog Number: (AZLOTWR038)
Supplier:  SCILABWARE
Description:   This stand is suitable for most round bottomed flasks.
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Holders made from PP for round bottom flasks.
1 - 8  of 11
  1  2  Next