Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Spectacles"
https://tr.vwr.com/stibo/smallweb/std.lang.all/53/50/18945350.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"154"
Sort by:
Catalog Number: (111-1846)
Supplier:  VWR Collection
Description:   The economical and compact cover glasses (OTG) fit over most prescription eyewear.
UOM:  1 * 1 items
Catalog Number: (111-1847)
Supplier:  VWR Collection
Description:   Wrap-around protective eyewear combining fashion, fit and function. One piece PC lens and a lightweight, frameless design ensures a comfortable and cu...
UOM:  1 * 1 items
Catalog Number: (111-0930)
Supplier:  VWR Collection
Description:   Visitor glasses with wrap-around clear polycarbonate lens and with indirect ventilation.
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   Lens cleaning tissue, grade 541, VWR®, Size: 80×100 mm
Supplier:  GE Healthcare - Whatman
Description:   These cleaning tissues are suitable for use with lenses and any other optical surfaces made from glass, quartz or plastic.
Catalog Number: (111-8030)
Supplier:  UVEX
Description:   Cover glasses (OTG) with an unrestricted lateral field of vision can be worn over most prescription spectacles.
UOM:  1 * 1 items
Supplier:  UVEX
Description:   Super OTG cover spectacles have thin, hingeless side arms with Grilamid® (a combination of hard and soft plastics). They combine the benefits of templ...
Supplier:  UVEX
Description:   The 100% metal free uvex i-works encapsulates the best of uvex eyewear: the sporty design, panoramic lens and low weight make it extremely comfortable...
1 - 8  of 154
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next