  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Specimen Jars"
https://tr.vwr.com/stibo/smallweb/std.lang.all/96/57/18709657.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
Sort by:
Supplier:  DURAN GROUP GMBH
Description:   Jars for specimen, DURAN® borosilicate glass, with ground joint. Low profile.
Supplier:  DURAN GROUP GMBH
Description:   Cylinder with knobbed lid, DURAN® Borosilicate glass, Capacity: 250 ml, Height: 80 mm, Ø ext.: 80 mm
Supplier:  DURAN GROUP GMBH
Description:   Specimen jar, DURAN® Borosilicate glass, Capacity: 750 ml, Height: 220 mm, Ø ext.: 90 mm
Catalog Number: (SCOT242042409)
Supplier:  DURAN GROUP GMBH
Description:   Organ storage jars, DURAN® Borosilicate glass, Capacity: 75 ml, Height: 75 mm, Ø ext.: 54 mm
UOM:  1 * 10 items
Supplier:  GLASWARENFABRIK KARL HECHT
Description:   Jars, made of glass or stainless steel, with base, without lids.
Supplier:  GLASWARENFABRIK KARL HECHT
Description:   Cylinder, Glass, Height: 200 mm, Ø ext.: 80 mm
1 - 6  of 6