  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Draining Racks"
https://tr.vwr.com/stibo/smallweb/std.lang.all/68/88/20236888.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"13"
Sort by:
Catalog Number: (211-0196)
Supplier:  VWR Collection
Description:   Rack, Draining, VWR®, Pegs: 90, Wire, vinyl coated, Drying rack, 457×181×525 mm
UOM:  1 * 1 items
Supplier:  BÜRKLE
Description:   Rack, Draining, PVC, Flexible, PVC, with PP rods 60 mm (15), 100 mm (56) and 150 mm (6), W×H: 600×600 mm
Supplier:  BÜRKLE
Description:   Rack, Draining without drainage channel, PVC, With 15 rods (60 mm), 56 rods (100 mm), 6 rods (150 mm), W×H: 600×600 mm
Supplier:  BÜRKLE
Description:   Rack, Draining, Rods, PP, Ø×L: 6×60 mm
Catalog Number: (149-1310)
Supplier:  KARTELL
Description:   Rack, Draining, Pegs: 72, PS, Draining rack, 450×110×630 mm
UOM:  1 * 1 items
Catalog Number: (BRND95105)
Supplier:  Brand
Description:   PS, with integral draining trough.
UOM:  1 * 1 items
Supplier:  KARTELL
Description:   Rack, Draining, Rods for narrow-necked vessels
UOM:  1 * 11 items
Supplier:  Brand
Description:   Rack, Draining, Pegs for draining rack, PS, for narrow mouth vessels, test tubes, small volumetric flasks and graduated cylinders
UOM:  1 * 11 items
1 - 8  of 13
  1  2  Next