  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Hydrometers"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/53/17/18945317.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"116"
Sort by:
Supplier:  VWR Collection
Description:   M100 hydrometers made of glass.
Supplier:  VWR Collection
Description:   Glass hydrometers (without thermometer).
Supplier:  VWR Collection
Description:   Safety sugar scale combined form thermo-hydrometers designed in the food and beverage and other industries to measure sucrose.
Supplier:  VWR Collection
Description:   Made from optical quality polycarbonate, these plain form hydrometers are designed for applications where rough handling or contamination could be an ...
Supplier:  ALLA ATELIERS
Description:   Hydrometers for low and high surface tension suitable for general purpose applications.
Supplier:  ALLA ATELIERS
Description:   These high precision alcoholometers with individual number on the scale and a calibration temperature of 20 °C.
Supplier:  ALLA ATELIERS
Description:   Baumé hydrometers calibrated at 15 °C.
Supplier:  ALLA ATELIERS
Description:   Hydrometers for the determination of sugar in alcohol.
1 - 8  of 116
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next