  • Criteria
    • In Stock (4)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Syringe Filters"
https://tr.vwr.com/stibo/smallweb/std.lang.all/98/82/18989882.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"38"
Sort by:
Catalog Number: (548-1072)
Supplier:  VWR Collection
Description:   Glass fibre prefilter with PP housing
UOM:  1 * 1.000 items
Supplier:  VWR Collection
Description:   Available with either PP, PTFE or nylon membrane. Membranes are available in either 0,2 or 0,45 µm pore size with the choice often made on analytical ...
Supplier:  VWR Collection
Description:   Available with either Cellulose Acetate or PES membrane. Sterile membranes with 0,2 µm pore size are ideal for sterile filtration applications and tho...
Supplier:  PALL LIFE SCIENCES
Description:   Syringe filter, Acrodisc®, PP, Membrane: PES, Non sterile, Pore size: 0,1 µm, Filtration area: 2,8 cm², Ø: 25 mm
Supplier:  PALL LIFE SCIENCES
Description:   Syringe filter, Acrodisc®, PP, Membrane: Borosilicate glass fibre, Non sterile, Pore size: 1,0 ** µm, * in AutoPack™ tubes, Filtration area: 2,8 cm², Ø: 25 mm
Supplier:  PALL LIFE SCIENCES
Description:   O-ring kit, For: AcroPrep™ 24 filtration system
Supplier:  PALL LIFE SCIENCES
Description:   Simplifies scale up and minimises revalidation; no need to change membrane materials during transition to pilot or production. PP housing; four membra...
Supplier:  PALL LIFE SCIENCES
Description:   Syringe filter, Acrodisc®, PP, Membrane: PS, Non sterile, Pore size: 0,2 µm, Filtration area: 2,8 cm², Ø: 25 mm
1 - 8  of 38
  1  2  3  4  5  Next