  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Filter Crucibles"
https://tr.vwr.com/stibo/smallweb/std.lang.all/52/26/18945226.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
Sort by:
Supplier:  VWR Collection
Description:   Porcelain, with porous bottom
Supplier:  ROBU GLASFILTER
Description:   Filtration crucibles made from borosilicate glass 3.3 with integrated sintered glass filter plate.
Supplier:  ROBU GLASFILTER
Description:   Filter crucible, Borosilicate glass 3.3, Porosity: 0, Capacity: 50 ml, Plate Ø: 30 mm
Supplier:  ROBU GLASFILTER
Description:   Filter crucible, Borosilicate glass 3.3, Porosity: 1, Capacity: 30 ml, Plate Ø: 34 mm
Catalog Number: (LENZ07185430)
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Filter crucible, Borosilicate glass 3.3, Porosity: 4, Capacity: 30 ml, Plate Ø: 30 mm
UOM:  1 * 1 items
Supplier:  MORGAN ADVANCED MATERIALS HALD
Description:   Filter crucible, Porcelain, Capacity: 25 ml, Plate Ø: 24 mm
Supplier:  DURAN GROUP GMBH
Description:   Filter crucible, DURAN® Borosilicate glass, Porosity: 1, Capacity: 15 ml, Plate Ø: 28 mm
Supplier:  SCILABWARE
Description:   These filtration crucibles are made from Pyrex® borosilicate glass.
1 - 8  of 8