Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Shoe and Boot Covers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/97/91/18989791.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"52"
Sort by:
Supplier:  VWR Collection
Description:   Non woven PP with a CPE laminated sole.
Supplier:  VWR Collection
Description:   Shoe covers made from 65% Low Density PE (LDPE) and 35% Linear Low Density PE (LLDPE).
Catalog Number: (113-8280)
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP.
UOM:  1 * 100 items
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP. With latex pattern.
UOM:  1 * 100 items
Catalog Number: (113-8287)
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP.
UOM:  1 * 300 items
Supplier:  VWR Collection
Description:   VWR Maximum SF boot covers are the cleanest and most durable within the entire VWR clothing line. Made from spunbonded polypropylene fabric (SPP) with...
Supplier:  VWR Collection
Description:   VWR Maximum SF shoe covers are the cleanest and most durable within the entire VWR clothing line. Made from spunbonded polypropylene fabric (SPP) coat...
Supplier:  VWR Collection
Description:   VWR Maximum SF shoe covers are the cleanest and most durable within the entire VWR clothing line. Made from spunbonded polypropylene fabric (SPP) wit...
1 - 8  of 52
  1  2  3  4  5  6  7  Next