Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Hoods"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/94/02/18989402.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"15"
Sort by:
Catalog Number: (113-8202)
Supplier:  VWR Collection
Description:   These poypropylene head and face covers are designed to protect users from potential environmental contaminations.
UOM:  1 * 500 items
Supplier:  3M
Description:   Replacement hood, Versaflo™, S-607
Supplier:  DUPONT PROTECTIVE APPAREL
Description:   This hood is made of Tyvek®, a flash spun high density polyethylene. The fabric is permeable to both air and water vapour, yet repels water-based liqu...
UOM:  1 * 100 items
Supplier:  DUPONT PROTECTIVE APPAREL
Description:   The hood is made of Tyvek® which is a fabric made of high density polyethylene using a patented flash spinning process. Tyvek® provides an inherent ba...
UOM:  1 * 100 items
Supplier:  DUPONT PROTECTIVE APPAREL
Description:   This hood is made of Tyvek®, a flash spun high density polyethylene. The fabric is permeable to both air and water vapour, yet repels water-based liqu...
UOM:  1 * 100 items
Supplier:  KIMBERLY CLARK
Description:   These hoods are made of breathable SMS fabric that adds a cloth-like feel. They are available in a standard design or with an integrated face mask.
Supplier:  NITRITEX
Description:   These disposable hoods are made of Clean-tough™ fabric which is spunbonded non woven PP laminated with a film of PE. The material provides protection ...
Supplier:  NITRITEX
Description:   These cleanroom hoods are made of MT031 fabric consisting of 99% polyester and 1% carbon.
1 - 8  of 15
  1  2  Next