  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Weighing Boats"
https://tr.vwr.com/stibo/smallweb/std.lang.all/83/80/18368380.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"32"
Sort by:
Supplier:  VWR Collection
Description:   Dishes, weighing, micro, oval, VWR®, 1,12 ml, Aluminium, Shape: Oval, 25×7×7 mm, I-Ø×H: - mm
Supplier:  VWR Collection
Description:   Weighing boat, disposable, VWR®, 7 ml, PS, Anti-static: -, Shape: Square, 46×46×8 mm, White
Supplier:  VWR Collection
Description:   Dishes and pans, made of aluminium.
Supplier:  VWR Collection
Description:   Dish, weighing, disposable, with tab, VWR®, 60 ml, Aluminium, Shape: Round, I-Ø×H: 57×16 mm
Supplier:  VWR Collection
Description:   Disposable weighing boats, VWR®, 25 ml, PS, Anti-static: -, Shape: Diamond, 80×50×14 mm, White
Supplier:  VWR Collection
Description:   Weighing dish, disposable, VWR®, 75 ml, Aluminium, Shape: Round, I-Ø×H: 65×30 mm
Supplier:  VWR Collection
Description:   Weighing boat, disposable, With pour spout, VWR®, PS, Anti-static: -, Shape: Flat bottom, Medium, 83×132×26 mm, White
Supplier:  VWR Collection
Description:   VWR® paper weighing boats are a first in eco friendly laboratory consumables.
1 - 8  of 32
  1  2  3  4  Next