  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Blood Lancets"
https://tr.vwr.com/stibo/smallweb/std.lang.all/49/54/18944954.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"5"
Sort by:
Catalog Number: (233-5351)
Supplier:  BRAUN B. (INC. PETZOLD)
Description:   Stainless steel, rust-proof
UOM:  1 * 200 items
Supplier:  MARIENFELD
Description:   Safety lancets with gamma-sterilised needle which remains fully secured in its casing – before and after the penetration. This prevents injuries or in...
Catalog Number: (MARI7300200)
Supplier:  MARIENFELD
Description:   Made of stainless steel of high grade quality X6 Cr 17 (in compliance with DIN EN 10088-2), chrome content 16 to 18 %.
UOM:  1 * 200 items
Catalog Number: (612-3747)
Supplier:  GLASWARENFABRIK KARL HECHT
Description:   For blood sampling, sterile packed.
UOM:  1 * 200 items
Supplier:  GREINER BIO ONE
Description:   Safety blood lancet, MiniCollect®, blade, MiniCollect®, Pink, Size: 1,0 mm
1 - 5  of 5