  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Blood Collection Tubes"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/20/18989320.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"11"
Sort by:
Catalog Number: (SLIN39375T)
Supplier:  STERILIN
Description:   Blood collection tube, Sterilin®, Volume: 5 ml, Cap colour: White
UOM:  1 * 420 items
Catalog Number: (720-0085)
Supplier:  STERILIN
Description:   These tubes are made up of PS or PP
UOM:  1 * 1.000 items
Supplier:  KIMA SAS DI CHIARIN RENZO & C
Description:   Blood collection tube, vacuum, Vacutest®, Volume: 2 ml, With: K2EDTA anticoagulant, Length: 75 mm, Ø: 13 mm, Cap colour: Pale purple
UOM:  1 * 1.000 items
Supplier:  GREINER BIO ONE
Description:   Blood collection tubes with EDTA, VACUETTE®, Volume: 1 ml, With: additive K3E K3EDTA, paediatric draw, Length: 75 mm, Ø: 13 mm, PET, Cap colour: Lavender
Supplier:  GREINER BIO ONE
Description:   Glucose determination tubes, VACUETTE®, Volume: 2 ml, With: additive sodium fluoride(FE)/K3EDTA, Length: 75 mm, Ø: 13 mm, PET, Cap colour: Grey
Supplier:  GREINER BIO ONE
Description:   Serum collecting tubes, VACUETTE®, Volume: 4,5 ml, With: additive Z serum clot activator and black ring, Length: 75 mm, Ø: 13 mm, PET, Cap colour: Red
Supplier:  GREINER BIO ONE
Description:   Plasma collecting tube, VACUETTE®, Volume: 2 ml, With: additive lithium heparin (LH), Length: 75 mm, Ø: 13 mm, PET, Cap colour: Green
Supplier:  GREINER BIO ONE
Description:   The tubes contain sodium citrate solutions in concentrations of 3.2% and are used for assays in haemostaseology. The mixing of 9 parts blood to 1 part...
1 - 8  of 11
  1  2  Next