  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "molecular-biology-stains"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/53/29/18945329.jpg
SearchPresentationType-HORIZONTAL
SearchResultCount:"10"
Sort by:
Supplier:  VWR Chemicals
Description:   Quick-View™ Fluorescent Viability Stain is a ready to use staining reagent for easy discrimination between live and dead mammalian cells. Supplied in ...
UOM:  1 * 5 ml
Catalog Number: (N734-5ML)
Supplier:  VWR Chemicals
Description:   Supplied as a 200X concentrate, Gold-N-Gel™ RNA dye is a non mutagenic, fluorescent in-gel stain for immediate visualisation of RNA bands on denaturin...
UOM:  1 * 5 ml
Supplier:  VWR Chemicals
Description:   Liquid Stable Nuclear Fast Red is a room temperature, stable solution formulated specifically to prevent precipitation during storage. Liquid-Stable N...
Supplier:  VWR Chemicals
Description:   Ready to use sensitive substrate for the detection of horseradish peroxidase (HRP) activity. Absorbs at 450 nm (yellow end-product). Ideal for ELISA a...
UOM:  1 * 100 ml
Catalog Number: (J239-5ML)
Supplier:  VWR Chemicals
Description:   Magenta loading dye, 10X
UOM:  1 * 5 ml
Catalog Number: (J644-100ML)
Supplier:  VWR Chemicals
Description:   Ready-to-use sensitive substrate for the detection of horseradish peroxidase activity. Absorbs at 450 nm (yellow end-product). Ideal for ELISA and sol...
UOM:  1 * 100 ml
SDS
Supplier:  VWR Chemicals
Description:   TMB (3,3',5,5'-tetramethylbenzidine) Membrane Substrate consists of two solutions that are mixed immediately before use for the detection of horseradi...
UOM:  1 * 100 ml
Supplier:  BIOTIUM INC.
Description:   PMA™ is a high affinity photoreactive DNA binding dye developed by Biotium. The dye alone is weakly fluorescent, but increases in fluorescence upon bi...
1 - 8  of 10
  1  2  Next