  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Histology Stains"
https://tr.vwr.com/stibo/smallweb/std.lang.all/53/15/18945315.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"9"
Sort by:
Supplier:  Merck
Description:   Dye solutions and staining kits for histology and cytology.
UOM:  1 * 400 Tests
Supplier:  Merck
Description:   Hard tissue like bones or teeth and all specimens of calcified tissue like bone marrow punches require special pretreatment before routine processing ...
UOM:  1 * 1 l
Supplier:  Merck
Description:   Cytological staining developed by Papanicolaou is still the standard technique for carcinoma and hormonal cycle diagnostics. Cytocolor® is a staining ...
Supplier:  VWR Chemicals
Description:   Contains a mixture of hydrochloric acid and alcohols with some EDTA. Ready to use for applications in histology and pathology laboratories. Recommende...
Supplier:  VWR Collection
Description:   Contains a mixture of formic acids and formalin. Ready to use for applications in histology and pathology laboratories. Recommended volume: 20x the fr...
SDS
Supplier:  VWR Collection
Description:   Vials with chinese ink that can be used in macroscopy as marking ink.
SDS
Supplier:  VWR Collection
Description:   Contains a mixture of hydrochloric acids. Ready to use for applications in histology and pathology laboratories. Recommended volume: 20x the fragment...
SDS
Supplier:  BIOTIUM INC.
Description:   High purity dyes for live/dead cell distinction.
UOM:  1 * 10 mg
1 - 8  of 9
  1  2  Next