  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Haematology Stains"
https://tr.vwr.com/stibo/smallweb/std.lang.all/40/50/18374050.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"5"
Sort by:
Supplier:  Merck
Description:   Staining solutions and kits for haematology
SDS
Catalog Number: (1.15924.0025)
Supplier:  Merck
Description:   Stains and dyes for microscopy
UOM:  1 * 25 g
Supplier:  VWR Chemicals
Description:   Staining solutions
Supplier:  VWR Chemicals
Description:   Staining solutions and kits for microscopy
Supplier:  VWR Chemicals
Description:   Special buffer tablets for haematology
SDS
1 - 5  of 5