  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Floating Tube Racks"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/95/71/18709571.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"4"
Sort by:
Supplier:  IKA (JANKE & KUNKEL )
Description:   Floating tube rack for 8×15 ml tubes
Supplier:  Bel-Art
Description:   Prep-Safe™ microcentrifuge tube floating rack, PP, with vortexing attachment, 12 place, neon yellow
Catalog Number: (BELAF188760020)
Supplier:  Bel-Art
Description:   Cyrotube floating racks, 20 places, PP, Ø 95 mm
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Prep-Safe™ microcentrifuge tube floating rack, natural, 28 place, PP
UOM:  1 * 1 items
1 - 4  of 4