  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Dairy Testing Bottles"
https://tr.vwr.com/stibo/smallweb/std.lang.all/68/76/20236876.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Supplier:  GERBER FUNKE DR N.
Description:   Babcock bottle, for milk, 0 - 8 %
Catalog Number: (614-2055)
Supplier:  GERBER FUNKE DR N.
Description:   Milk sample bottles, PP with metal bottom, 50 ml capacity
UOM:  1 * 100 items
1 - 2  of 2