  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Slide Warmers/Dryers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/98/05/18989805.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Catalog Number: (720-2422)
Supplier:  VWR Collection
Description:   The W10 heating plate with its sleek, compact design accommodates two rows of up to 40 slides in total. The heating plate is very useful as a stretch ...
UOM:  1 * 1 items
Supplier:  ELECTROTHERMAL ENGIN.
Description:   The bench has a capacity of fifty 26×76 mm slides, which are placed directly on the warming plate or on the adjustable support rods, either horizontal...
1 - 2  of 2