Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Reagents"
https://tr.vwr.com/stibo/smallweb/std.lang.all/97/16/18989716.jpg
SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"7"
Sort by:
Supplier:  VWR Chemicals
Description:   Bis-Tris hydrochloride
Supplier:  VWR Chemicals
Description:   DNA, salt, fish sperm
Supplier:  VWR Chemicals
Description:   Luciferin, sodium salt ultra pure grade
Catalog Number: (1B1524-500G)
Supplier:  VWR Chemicals
Description:   Pluronic® PE 6800 reagent grade
UOM:  1 * 500 g
Supplier:  VWR Chemicals
Description:   Brij O10
Catalog Number: (786-293)
Supplier:  G-Biosciences
Description:   Molecular biology reagents
UOM:  1 * 1 l
Catalog Number: (J60342.14)
Supplier:  ALFA (AESAR)
Description:   Clofibrate
UOM:  1 * 25 g
SDS
1 - 7  of 7