  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Ampoules"
https://tr.vwr.com/stibo/smallweb/std.lang.all/92/92/18989292.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Ampoule, pre-scored, amber, Borosilicate glass, 2 ml, Ø×H: 12×75 mm
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Ampoule, standard, Borosilicate glass, 20 ml, Ø×H: 28×100 mm
Catalog Number: (WHEAW651446)
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Lyophilisation ampoule, stopper sealed, Vacule®, Borosilicate glass, 2 ml, Ø×H: 12×86 mm, Ø ext. top: 10 mm
UOM:  1 * 567 items
Supplier:  DWK Life Sciences (WHEATON®)
Description:   Lyophilisation ampoule, pre-scored, Vacule®, Borosilicate glass, 2 ml, Ø×H: 11,8×122 mm, Ø ext. top: 11,75 mm
Supplier:  DWK Life Sciences (WHEATON®)
Description:   All glass ampoules that can be hermetically sealed to preserve sample purity and extend sample shelf life. Ideal for standards packaging, environmenta...
1 - 5  of 5