  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Ductless Fume Hoods"
https://tr.vwr.com/stibo/smallweb/std.lang.all/51/61/18945161.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"15"
Sort by:
Catalog Number: (ERLAI2820000003)
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Mobile ductless filtration fume hood for personal protection during handlings of liquid, gaseous and solid chemicals. Molecular filtration by adsorpti...
UOM:  1 * 1 items
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Ductless fume hood, captair®, Captair® Smart 321 1C with one carbon filter GF4/F (for formaldehyde vapours), Material: Galvanized Steel, W×D×H ext.: 765×615×1285 mm, W×D×H int.: 700×533×1100 mm, Lighting: LED > 650 lux
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Ductless fume hood, captair®, Captair® Smart 391 1C with one carbon filter GF4/K (for ammonia vapours), Material: Galvanized Steel, W×D×H ext.: 980×615×1285 mm, W×D×H int.: 895×533×1100 mm, Lighting: LED > 650 lux
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Ductless fume hood, captair®, Captair® Smart 481 1P1C with one HEPA and one carbon filter GF4/F (for formaldehyde vapours), Material: Galvanized Steel, W×D×H ext.: 1245×615×1285 mm, W×D×H int.: 1176×533×1100 mm, Lighting: LED > 650 lux
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   Ceiling-mounted laboratory-grade filtration unit.
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive...
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive...
Supplier:  ERLAB (INCL. CAPTAIRE)
Description:   The new Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive...
1 - 8  of 15
  1  2  Next