Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Distillation Components"
https://tr.vwr.com/stibo/smallweb/std.lang.all/51/38/18945138.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"87"
Sort by:
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Measuring tube with stopcock (Zero point at stopcock), Cone size: 29/32 NS, Socket size: 29/32 NS
Supplier:  WHEATON INDUSTRIES INC
Description:   Distillation apparatus, with grease free, screw-threaded Wheaton Connection®, Thermometer adapter, 20-400
UOM:  1 * 1 items
Supplier:  WHEATON INDUSTRIES INC
Description:   Distillation apparatus, with standard taper 19/22 Clear-Seal® joints, Borosilicate glass, Connecting adapter
Supplier:  WHEATON INDUSTRIES INC
Description:   Cyanide distillation kit with Clear-Seal® joints, Borosilicate glass, Cyanide distillation kit complete
UOM:  1 * 1 items
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Bends, with two cones, 75°, bent, DURAN® Borosilicate glass, Cone size: 14/23 NS
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Adapters, delivery, bent, 105°, with short tube, DURAN® Borosilicate glass, Socket size: 19/26 NS, Tube length: 65 mm
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Distilling Links, with Liebig condenser, one thermometer socket, and water connection with glass olives, DURAN® Borosilicate glass, Jacket length: 400 mm, Cone size: 29/32 NS, Socket size: 14/23 NS
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Distilling links, Claisen, with liebig condenser, vacuum receiver adapter and GL thread, DURAN® Borosilicate glass, Jacket length: 250 mm, Cone size: 14/23 NS, Socket size: 14/23 NS
1 - 8  of 87
  1  2  3  4  5  6  7  8  9  10  11  Next