Would you like to log in again?
  • Product Category
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Ice Containers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/94/19/18989419.jpg
SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
SearchResultCount:"12"
Sort by:
Supplier:  VWR Collection
Description:   Polyurethane foam.
Supplier:  VWR Collection
Description:   Polyurethane foam.
Catalog Number: (BCISBCS-111PL)
Supplier:  BIOCISION, LLC
Description:   Ice pan, rectangular, Maxi, PVC, Capacity: 9 l, Colour: Purple, W×D×H: 406×318×114 mm
UOM:  1 * 1 items
Discontinued
Supplier:  Corning Life Science
Description:   Capacity: 4 l, Colour: Green
Catalog Number: (KART1000)
Supplier:  KARTELL
Description:   Made of red PUR. Equipped with lid and two side-handles. Can be used with dry or minced ice and cooling mixtures.
UOM:  1 * 1 items
Catalog Number: (BRND156100)
Supplier:  Brand
Description:   Ice bucket, octagonal, with lid, PU, Capacity: 4,5 l, W×D×H: 280×330×180 mm
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Ice bucket, Magic Touch™, round, with lid, Urethane, Capacity: 2,5 l, Ø ext.: 268 mm, Height: 179 mm, Colour: Black
Supplier:  Bel-Art
Description:   Ice pan, Mini, with lid, Magic Touch™, Urethane, Capacity: 1 l, Colour: Black, W×D×H: 183×183×135 mm
1 - 8  of 12
  1  2  Next