  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Desiccators, Glass"
https://tr.vwr.com/stibo/smallweb/std.lang.all/95/48/18709548.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"14"
Sort by:
Supplier:  VWR Collection
Description:   Desiccator, VWR®, Borosilicate glass 3.3, With stopcock, Nominal size: 100, Ø ext.×H: 153×180 mm, Ø int.: 92 mm
Supplier:  VWR Collection
Description:   Desiccator, Borosilicate glass 3.3, Nominal size: 250 DN, Usable height: 140 mm, Ø ext.×H: 320×370 mm, Ø int.: 239 mm
Supplier:  DWK Life Sciences (DURAN®)
Description:   Desiccator, DURAN®, Borosilicate glass, Mobilex, Nominal size: 300 DN, Usable height: 150 mm, Ø ext.×H: 380×420 mm
Supplier:  DWK Life Sciences (DURAN®)
Description:   Clear borosilicate glass. Choice of models with flat flange, NOVUS NS 24/29 junction, and outlet in lid with stopcock or desiccators with flat flange ...
Supplier:  DWK Life Sciences (DURAN®)
Description:   Clear borosilicate glass 3.3, with Mobilex screw-thread in base and lid with stopcock.
Supplier:  Brand
Description:   Borosilicate glass 3.3 with precisely ground flat flanges.
Supplier:  Brand
Description:   Borosilicate glass 3.3 with precisely ground flat flanges.
Supplier:  WITEG LABORTECHNIK GMBH
Description:   Desiccators are made of borosilicate glass, with safety PROTEFAN coating as implosion and splinter protection.
1 - 8  of 14
  1  2  Next